ENA429359258 Enamine

1-{2-amino-5H,6H,7H,8H-pyrido[4,3-d]pyrimidin-6-yl}ethan-1-one

  • MDL number MFCD11655916
  • Cas Number : 1100752-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187393

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187393
Availability details for
Quantity
  • Availability
  • Add to Cart