ENA429359261 Enamine

5-bromo-2-methyl-3-[(4-methyl-1,2,5-oxadiazol-3-yl)methyl]-3,4-dihydropyrimidin-4-one

  • MDL number MFCD22801767
  • Cas Number : 1513494-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187398

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187398
Availability details for
Quantity
  • Availability
  • Add to Cart