ENA429359267 Enamine

2-amino-3-cyclopropylpropan-1-ol hydrochloride

  • MDL number MFCD08455506
  • Cas Number : 1379954-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187426

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187426
Availability details for
Quantity
  • Availability
  • Add to Cart