ENA429359269 Enamine

1-[3-(4-methylpyridin-2-yl)-1H-1,2,4-triazol-5-yl]ethan-1-amine hydrochloride

  • MDL number MFCD28118638
  • Cas Number : 1798732-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187428

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187428
Availability details for
Quantity
  • Availability
  • Add to Cart