ENA429359276 Enamine

tert-butyl 2-methylpent-4-enoate

  • MDL number MFCD26402144
  • Cas Number : 93958-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187461

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187461
Availability details for
Quantity
  • Availability
  • Add to Cart