ENA429359278 Enamine

4-benzylpiperidine-1-carbonyl chloride

  • MDL number MFCD19281929
  • Cas Number : 31252-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187469

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187469
Availability details for
Quantity
  • Availability
  • Add to Cart