ENA429359279 Enamine

ethyl 3-(1-amino-3-methylbutyl)-1H-1,2,4-triazole-5-carboxylate

  • MDL number MFCD21741007
  • Cas Number : 1539914-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187470
Availability details for
Quantity
  • Availability
  • Add to Cart