ENA429359281 Enamine

1-(3-aminophenyl)-1-methylurea

  • MDL number MFCD14617411
  • Cas Number : 1247579-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187473
Availability details for
Quantity
  • Availability
  • Add to Cart