ENA429359284 Enamine

{[1-(2,2-difluoroethyl)-1H-1,2,4-triazol-5-yl]methyl}(methyl)amine

  • MDL number MFCD21272441
  • Cas Number : 1497455-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187476
Availability details for
Quantity
  • Availability
  • Add to Cart