ENA429359285 Enamine

[(2S)-1-[(3-fluorophenyl)methyl]pyrrolidin-2-yl]methanol

  • MDL number MFCD21644674
  • Cas Number : 892386-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187477
Availability details for
Quantity
  • Availability
  • Add to Cart