ENA429359286 Enamine

2,3-dichloropyridine-4-carbonitrile

  • MDL number MFCD13193096
  • Cas Number : 184416-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187478
Availability details for
Quantity
  • Availability
  • Add to Cart