ENA429359288 Enamine

3-(3-iodophenoxy)propanoic acid

  • MDL number MFCD11182861
  • Cas Number : 1094426-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187481
Availability details for
Quantity
  • Availability
  • Add to Cart