ENA429359292 Enamine

(5-amino-2-methoxyphenyl)urea

  • MDL number MFCD09049155
  • Cas Number : 926205-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187488

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187488
Availability details for
Quantity
  • Availability
  • Add to Cart