ENA429359293 Enamine

2-carbamothioyl-N-[2-(3,4-dimethoxyphenyl)ethyl]acetamide

  • MDL number MFCD12559497
  • Cas Number : 1038973-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187491
Availability details for
Quantity
  • Availability
  • Add to Cart