ENA429359294 Enamine

2-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD11650804
  • Cas Number : 1099610-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187492
Availability details for
Quantity
  • Availability
  • Add to Cart