ENA429359295 Enamine

1-[(4-chlorophenyl)methyl]-3-(furan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD18940428
  • Cas Number : 1408705-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187493

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187493
Availability details for
Quantity
  • Availability
  • Add to Cart