ENA429359297 Enamine

methyl 4-(oxolan-3-yloxy)benzoate

  • MDL number MFCD21120461
  • Cas Number : 1497119-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187496
Availability details for
Quantity
  • Availability
  • Add to Cart