ENA429359302 Enamine

2-(2-bromoethyl)phenol

  • MDL number MFCD09744987
  • Cas Number : 57027-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187505

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187505
Availability details for
Quantity
  • Availability
  • Add to Cart