ENA429359306 Enamine

1,2,3,4-tetrahydronaphthalen-2-ylmethanol

  • MDL number MFCD09868527
  • Cas Number : 6947-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187509

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187509
Availability details for
Quantity
  • Availability
  • Add to Cart