ENA429359307 Enamine

[5-(3-fluorophenyl)-1,2-oxazol-3-yl]methanol

  • MDL number MFCD17179176
  • Cas Number : 1378817-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187512

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187512
Availability details for
Quantity
  • Availability
  • Add to Cart