ENA429359310 Enamine

1-(3-amino-2-methylphenyl)imidazolidin-2-one

  • MDL number MFCD11167825
  • Cas Number : 1042555-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187515

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P305+P351+P338,P330,P337+P313,P501
MSDS https://www.enaminestore.com/catalog/EN300-187515
pictograms GHS07
hstatements H302,H319
Availability details for
Quantity
  • Availability
  • Add to Cart