ENA429359311 Enamine

1-(5-amino-2-methylphenyl)imidazolidin-2-one

  • MDL number MFCD11167827
  • Cas Number : 1038247-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187516
Availability details for
Quantity
  • Availability
  • Add to Cart