ENA429359312 Enamine

1-(3-amino-4-fluorophenyl)imidazolidin-2-one

  • MDL number MFCD11167821
  • Cas Number : 1042555-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187517
Availability details for
Quantity
  • Availability
  • Add to Cart