ENA429359314 Enamine

methyl 5-[(methylamino)methyl]furan-2-carboxylate hydrochloride

  • MDL number MFCD28118642
  • Cas Number : 1803588-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187519
Availability details for
Quantity
  • Availability
  • Add to Cart