ENA429359320 Enamine

5-chloro-4-methoxy-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD21874976
  • Cas Number : 1368747-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187538
Availability details for
Quantity
  • Availability
  • Add to Cart