ENA429359321 Enamine

4-chloro-3-(hydroxymethyl)phenol

  • MDL number MFCD09038291
  • Cas Number : 876299-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187542
Availability details for
Quantity
  • Availability
  • Add to Cart