ENA429359325 Enamine

4-(morpholine-4-carbonyl)-1H-pyrazol-5-amine

  • MDL number MFCD11105378
  • Cas Number : 1239694-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187551
Availability details for
Quantity
  • Availability
  • Add to Cart