ENA429359330 Enamine

(2-fluoro-4,6-dimethylphenyl)methanol

  • MDL number MFCD18825450
  • Cas Number : 252004-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187567

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187567
Availability details for
Quantity
  • Availability
  • Add to Cart