ENA429359333 Enamine

5-chloroisoquinolin-8-amine

  • MDL number MFCD19687159
  • Cas Number : 55766-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187575

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187575
Availability details for
Quantity
  • Availability
  • Add to Cart