ENA429359339 Enamine

2-ethylbenzene-1-sulfonamide

  • MDL number MFCD11650798
  • Cas Number : 85-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187615
Availability details for
Quantity
  • Availability
  • Add to Cart