ENA429359341 Enamine

4-(difluoromethoxy)-2,5-difluoroaniline

  • MDL number MFCD18844219
  • Cas Number : 1341923-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187618

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187618
Availability details for
Quantity
  • Availability
  • Add to Cart