ENA429359342 Enamine

1-(methylsulfanyl)cyclobutane-1-carboxylic acid

  • MDL number MFCD19229084
  • Cas Number : 35120-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187619
Availability details for
Quantity
  • Availability
  • Add to Cart