ENA429359343 Enamine

1-(2,2-difluoroethyl)-2-ethyl-1H-imidazole

  • MDL number MFCD18889566
  • Cas Number : 1339790-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187620
Availability details for
Quantity
  • Availability
  • Add to Cart