ENA429359344 Enamine

4-methyl-1-(propan-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD11215416
  • Cas Number : 1174866-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187621
Availability details for
Quantity
  • Availability
  • Add to Cart