ENA429359346 Enamine

2-chloro-5-methanesulfonylpyrimidine

  • MDL number MFCD21879442
  • Cas Number : 321565-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187624

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187624
Availability details for
Quantity
  • Availability
  • Add to Cart