ENA429359347 Enamine

[4-(2-fluorophenyl)-1,3-thiazol-2-yl]methanamine

  • MDL number MFCD05222014
  • Cas Number : 643725-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187625

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187625
Availability details for
Quantity
  • Availability
  • Add to Cart