ENA429359350 Enamine

N,1-dimethylcyclopentan-1-amine hydrochloride

  • MDL number MFCD28118654
  • Cas Number : 89854-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187630
Availability details for
Quantity
  • Availability
  • Add to Cart