ENA429359354 Enamine

3-(methoxymethyl)pyridine-2-carbonitrile

  • MDL number MFCD17243360
  • Cas Number : 863548-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187638

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187638
Availability details for
Quantity
  • Availability
  • Add to Cart