ENA429359357 Enamine

(2,2-dimethylcyclopentyl)hydrazine hydrochloride

  • MDL number MFCD28118657
  • Cas Number : 1803608-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187641

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187641
Availability details for
Quantity
  • Availability
  • Add to Cart