ENA429359358 Enamine

4-[(piperidin-4-yl)methyl]pyridine

  • MDL number MFCD05189968
  • Cas Number : 899356-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187642
Availability details for
Quantity
  • Availability
  • Add to Cart