ENA429359359 Enamine

1-cyclopropylpyrrolidine-2-carboxylic acid hydrochloride

  • MDL number MFCD28125025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187644
Availability details for
Quantity
  • Availability
  • Add to Cart