ENA429359360 Enamine

4-(4-fluorophenyl)oxan-4-amine hydrochloride

  • MDL number MFCD22418720
  • Cas Number : 1385696-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187645
Availability details for
Quantity
  • Availability
  • Add to Cart