ENA429359361 Enamine

ethyl 3-(1H-indol-3-yl)butanoate

  • MDL number MFCD28118658
  • Cas Number : 67996-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187647
Availability details for
Quantity
  • Availability
  • Add to Cart