ENA429359362 Enamine

[1,3-dimethyl-5-(2-methyl-1,4-oxazepan-4-yl)-1H-pyrazol-4-yl]methanamine

  • MDL number MFCD17413503
  • Cas Number : 1275870-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187649

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187649
Availability details for
Quantity
  • Availability
  • Add to Cart