ENA429359363 Enamine

1-cyclopropyl-3,3,3-trifluoropropan-1-amine hydrochloride

  • MDL number MFCD28118659
  • Cas Number : 1788615-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187650
Availability details for
Quantity
  • Availability
  • Add to Cart