ENA429359365 Enamine

1-(3-acetylphenyl)imidazolidin-2-one

  • MDL number MFCD23745056
  • Cas Number : 1012040-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187652
Availability details for
Quantity
  • Availability
  • Add to Cart