ENA429359366 Enamine

1-bromo-2,4-difluoro-3-iodobenzene

  • MDL number MFCD12922602
  • Cas Number : 1208075-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187653

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187653
Availability details for
Quantity
  • Availability
  • Add to Cart