ENA429359372 Enamine

4-(piperidin-1-ylmethyl)benzoic acid hydrochloride

  • MDL number MFCD09759039
  • Cas Number : 106261-47-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187681

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187681
Availability details for
Quantity
  • Availability
  • Add to Cart