ENA429359382 Enamine

4-(2,2-dimethylpropanamido)cyclohexane-1-carboxylic acid

  • MDL number MFCD12156971
  • Cas Number : 1184664-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-187730

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187730
Availability details for
Quantity
  • Availability
  • Add to Cart