ENA429359383 Enamine

4-cyclopropaneamidocyclohexane-1-carboxylic acid

  • MDL number MFCD12156974
  • Cas Number : 1184684-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-187731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-187731
Availability details for
Quantity
  • Availability
  • Add to Cart